Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTG1 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI7H6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270286AF405

Catalog No. N270286A405


Add to cart

Description

Description

BTG1 Monoclonal antibody specifically detects BTG1 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

BTG1
OTI7H6
Immunohistochemistry
Affinity Purified, Conjugated
BTG1
Full length recombinant protein of human BTG1 (NP_001722) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
B-cell translocation gene 1 protein, B-cell translocation gene 1, proliferative, protein BTG1
Mouse
IgG1
0.1 mL
Primary
694.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit