Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTG1 Mouse anti-Human, DyLight 405, Clone: OTI7H6, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270286V
Description
BTG1 Monoclonal antibody specifically detects BTG1 in Human samples. It is validated for Immunohistochemistry.Specifications
BTG1 | |
OTI7H6 | |
Immunohistochemistry | |
Affinity Purified, Conjugated | |
BTG1 | |
Full length recombinant protein of human BTG1 (NP_001722) produced in E.coli. | |
Immunogen affinity purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Immunohistochemistry | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
B-cell translocation gene 1 protein, B-cell translocation gene 1, proliferative, protein BTG1 | |
Mouse | |
IgG1 | |
0.1 mL | |
Primary | |
694.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title