Learn More
BTG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148791AP
Catalog No. 50-172-9800
Add to cart
Description
Description
BTG1 is a member of an anti-proliferative gene family that regulates cell growth and differentiation. Expression of this gene is highest in the G0/G1 phases of the cell cycle and downregulated when cells progressed through G1. The encoded protein interacts with several nuclear receptors, and functions as a coactivator of cell differentiation. This locus has been shown to be involved in a t(8; 12)(q24; q22) chromosomal translocation in a case of B-cell chronic lymphocytic leukemia.
Specifications
Specifications
BTG1
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62324, P62325, Q63073
BTG1, Protein BTG1
Rabbit
IgG
150 μL
-20°C
Primary
12226, 29618, 694
Immunohistochemistry (Paraffin)
Unconjugated
BTG1
Liquid
BTG1
BTG1 Fusion Protein Ag6651
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat