Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTG2 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI4C10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271339AF647

Catalog No. N271339A647


Add to cart

Description

Description

BTG2 Monoclonal antibody specifically detects BTG2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

BTG2
OTI4C10
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
BTG2
Full length recombinant protein of human BTG2 (NP_006754) produced in E.coli.
Immunogen affinity purified
Cell Cycle and Replication
Primary
7832.0
Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
B-cell translocation gene 2, BTG family member 2, BTG family, member 2, MGC126063, MGC126064, NGF-inducible proliferative protein PC3, PC3nerve growth factor-inducible proliferative, pheochromacytoma cell-3, protein BTG2, TIS21
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit