Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTG2 Rabbit anti-Human, DyLight 405, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297919V
Description
BTG2 Polyclonal specifically detects BTG2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BTG2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
B-cell translocation gene 2, BTG family member 2, BTG family, member 2, MGC126063, MGC126064, NGF-inducible proliferative protein PC3, PC3nerve growth factor-inducible proliferative, pheochromacytoma cell-3, protein BTG2, TIS21 | |
Produced in rabbits immunized with E. coli-derived Human BTG2 fragment. | |
0.1 mL | |
Cell Cycle and Replication | |
7832 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title