Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTG2 Mouse anti-Human, Clone: OTI4C10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526949

Catalog No. PIMA526949


Add to cart

Description

Description

The protein encoded by this gene is a member of the BTG/Tob family. This family has structurally related proteins that appear to have antiproliferative properties. This encoded protein is involved in the regulation of the G1/S transition of the cell cycle.
Specifications

Specifications

BTG2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P78543
PC3, TIS21, B-cell translocation gene 2, NGF-inducible anti-proliferative protein PC3, nerve growth factor-inducible anti-proliferative, pheochromacytoma cell-3, protein BTG2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7832
Immunohistochemistry (Paraffin), Western Blot
OTI4C10
Unconjugated
BTG2
Liquid
BTG2
Full length human recombit protein of BTG2 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit