Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTG3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548438

Catalog No. PA548438

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Overexpression impairs serum-induced cell cycle progression from the G0/G1 to S phase.
Specifications

Specifications

BTG3
Polyclonal
PBS with 0.09% sodium azide
Q14201
Protein BTG3, Abundant in neuroepithelium area protein, BTG family member 3, Protein Tob5, BTG3, ANA, TOB5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10950
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
BTG3
Liquid
BTG3
KLH conjugated synthetic peptide between 100-129 amino acids from the Central region of human BTG3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit