Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTG4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549030

Catalog No. PA549030

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Shows marked antiproliferative activity, being able to induce G(1) arrest.
Specifications

Specifications

BTG4
Polyclonal
PBS with 0.09% sodium azide
Q9NY30
Protein BTG4, BTG family member 4, Protein PC3b, BTG4, PC3B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54766
Western Blot
Unconjugated
BTG4
Liquid
BTG4
KLH conjugated synthetic peptide between 142-169 amino acids from the C-terminal region of human BTG4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit