Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTK Mouse anti-Human, Clone: OTI10E10, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA501118S
Description
BTK (Bruton ft.s tyrosine kinase or tyrosine-protein kinase BTK) was initially identified as a member of the src family for protein-tyrosine kinases that was involved in X-linked a-gamma-globulinaemia, and has since been shown to be involved in a number of signaling pathways in hemapoietic lineage. It has recently been shown to interact with members of the toll-like receptor (TLR) family such as TLR4, 6, 8, and 9. The TLRs are critical molecules in both the innate and adaptive immunity and can recognize diverse microbial pathogens. BTK has also been shown to interact with key proteins involved in TLR4 signal transduction such as MyD88, TIRAP, and IRAK, but not TRAF-6, suggesting that BTK is involved in lipopolysaccharide signal transduction. Mutations in this gene cause X-linked agammaglobulinemia type 1, which is an immunodeficiency characterized by the failure to produce mature B lymphocytes, and associated with a failure of Ig heavy chain rearrangement.Specifications
BTK | |
Monoclonal | |
0.87 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q06187 | |
AGMX1, AT, ATK, BPK, IMD1, PSCTK1, XLA | |
Mouse | |
IgG2a | |
30 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
695 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI10E10 | |
Unconjugated | |
BTK | |
Liquid | |
BTK | |
Full length human recombit protein of human BTK produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title