Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTK Mouse anti-Human, PE/Atto594, Clone: OTI3A2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270291PEATT594

Catalog No. N270291P594


Add to cart

Description

Description

BTK Monoclonal antibody specifically detects BTK in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

BTK
OTI3A2
Flow Cytometry
Affinity Purified, Conjugated
BTK
Full length recombinant protein of human BTK (NP_000052) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis, Immunology, Innate Immunity, Phospho Specific
Primary
695.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit