Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human BTK, His Tag Recombinant Protein
BTK Recombinant Human Protein
$551.65 - $1853.65
Specifications
Accession Number | NP_000052 |
---|---|
Gene Alias | BTK, ATK, XLA |
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Description
Bruton tyrosine kinase (BTK) is a cytoplasmic tyrosine kinase belonging to the SRC-related TEC subfamily of tyrosine kinases. Mutations in the BTK gene have been linked to severe developmental blocks in human B-cell ontogeny and immunodeficiency disorders.Specifications
NP_000052 | |
Human | |
Baculovirus |
BTK, ATK, XLA | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title