Learn More
Andwin Scientific BTL SPRAY DISPENSER 22OZ 4/CS
SDP
Supplier:  Andwin Scientific 7340X
Encompass_Preferred
Adjustable Spray Bottle, Have a trigger-motion spray that dispenses a spray or stream of liquid from an adjustable nozzle, An attached strainer helps keep solid particles from clogging the sprayer mechanism, 22 oz., 660mL, 1/EA
Catalog No. NC9278158