Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTLA/CD272 Mouse anti-Human, Clone: BTLA/1528, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen BTLA/CD272
Clone BTLA/1528
Concentration 0.2 mg/ml
Dilution ELISA
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123079
View Documents
Novus Biologicals
NBP30780310UG
100 μg
Each for $352.89
Add to cart
 
NB123080
View Documents
Novus Biologicals
NBP30780320UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Human samples. It is validated for ELISA
Specifications

Specifications

BTLA/CD272
0.2 mg/ml
ELISA
Purified
RUO
Human
B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743
Recombinant full-length human BTLA/CD272 protein
Monoclonal
Protein A or G purified
BTLA/1528
ELISA
Unconjugated
Mouse
Cell Cycle and Replication, Phospho Specific
10 mM PBS with 0.05% BSA
151888
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit