Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTLA/CD272, Rabbit anti-Human, Clone: 2161A, R&D Systems™

Rabbit Monoclonal Antibody

$131.70 - $315.00

Specifications

Antigen BTLA/CD272
Clone 2161A
Immunogen Mouse myeloma cell line, NS0-derived human BTLA, Ile25-Ser150, Accession # AAP44003
Host Species Rabbit
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB33542100
View Documents
R&D Systems
MAB33542100
100 ug
Each for $315.00
Add to cart
 
MAB33542SP
View Documents
R&D Systems
MAB33542SP
25 ug
Each for $131.70
Add to cart
 
Description

Description

BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Human Samples. It is validated for Flow Cytometry, CyTOF.
Specifications

Specifications

BTLA/CD272
Mouse myeloma cell line, NS0-derived human BTLA, Ile25-Ser150, Accession # AAP44003
Primary
Human
Unconjugated
151888
Purified
Flow Cytometry, CyTOF
RUO
2161A
Rabbit
Monoclonal
IgG
B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit