Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTLA/CD272, Rabbit anti-Mouse, Clone: HMBT-6B2, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen BTLA/CD272
Clone HMBT-6B2
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay, Immunoblot, Blocking Assay
Host Species Rabbit
Target Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB016500
View Documents
Novus Biologicals
NBP2811180.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB016501
View Documents
Novus Biologicals
NBP2811180.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Mouse Samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Functional, Immunoblotting, Blocking Assay.
Specifications

Specifications

BTLA/CD272
Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay, Immunoblot, Blocking Assay
Mouse
Primary
HMBT-6B2
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit