Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTLA Mouse anti-Human, Clone: 4C2, Abnova™

Mouse monoclonal antibody raised against full length recombinant human BTLA.

Manufacturer:  Abnova Corporation MAB12997

Catalog No. 89-933-676


Add to cart

Description

Description

BTLA expression is induced during activation of T cells, and BTLA remains expressed on Th1 cells but not Th2 cells. Like PD1 (MIM 600244) and CTLA4 (MIM 123890), BTLA interacts with a B7 homolog, B7H4.[supplied by OMIM]

Specifications

Specifications

BTLA
4C2
Mouse monoclonal antibody raised against full length recombinant human BTLA.
In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
BTLA1/CD272/FLJ16065/MGC129743
Mouse
IgG2b
50 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Liquid
BTLA
Recombinant protein corresponding to full length human BTLA.
Protein A/G purification
RUO
Primary
151888
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit