Learn More
BTN1A1/Butyrophilin Antibody (OTI7B5), DyLight 550, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270299R
Catalog No. NBP270299R
Add to cart
Description
Description
BTN1A1/Butyrophilin Monoclonal specifically detects BTN1A1/Butyrophilin in Human, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
BTN1A1/Butyrophilin
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
BTN1A1
Full length recombinant protein of human BTN1A1/Butyrophilin (NP_001723) produced in HEK293T cells.
0.1 mL
696.0
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI7B5
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1
Mouse
Affinity Purified
Primary
Human, Canine
IgG2a
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'