Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTN1A1/Butyrophilin Mouse anti-Human, Canine, Allophycocyanin/Cy7, Clone: OTI7B5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270299APCCY7

Catalog No. N270299APC7


Add to cart

Description

Description

BTN1A1/Butyrophilin Monoclonal antibody specifically detects BTN1A1/Butyrophilin in Human, Canine samples. It is validated for Flow Cytometry.
Specifications

Specifications

BTN1A1/Butyrophilin
OTI7B5
Flow Cytometry
Affinity Purified, Conjugated
BTN1A1
Full length recombinant protein of human BTN1A1/Butyrophilin (NP_001723) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1
Mouse
IgG2a
0.1 mL
Primary
696.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit