Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTN1A1/Butyrophilin Mouse anti-Human, Canine, DyLight 755, Clone: OTI7B5, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270299IR
Description
BTN1A1/Butyrophilin Monoclonal antibody specifically detects BTN1A1/Butyrophilin in Human, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
BTN1A1/Butyrophilin | |
OTI7B5 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Affinity Purified, Conjugated | |
BTN1A1 | |
Full length recombinant protein of human BTN1A1/Butyrophilin (NP_001723) produced in HEK293T cells. | |
Immunogen affinity purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Canine, Human |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1 | |
Mouse | |
IgG2a | |
0.1 mL | |
Primary | |
696.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title