Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTN1A1 Mouse anti-Canine, Human, Clone: OTI7B5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525201

Catalog No. PIMA525201


Add to cart

Description

Description

May function in the secretion of milk-fat droplets. May act as a specific membrane-associated receptor for the association of cytoplasmic droplets with the apical plasma membrane Inhibits the proliferation of CD4 and CD8 T-cells activated by anti-CD3 antibodies, T-cell metabolism and IL2 and IFNG secretion
Specifications

Specifications

BTN1A1
Monoclonal
0.58 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q13410
BTN1A1; BTN
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100855656, 696
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI7B5
Unconjugated
BTN1A1
Liquid
BTN1A1
Full length human recombit protein of BTN1A1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit