Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTN2A1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen BTN2A1
Immunogen Synthetic peptides corresponding to BTN2A1(butyrophilin, subfamily 2, member A1) The peptide sequence was selected from the N terminal of BTN2A1. Peptide sequence SVALVIHNITAQENGTYRCYFQEGRSYDEAILHLVVAGLGSKPLISMRGH.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1623720 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16237120UL
20ul Each for $149.00
Add to cart
 
NBP162371 View Documents Novus BiologicalsSupplier Diversity Partner
NBP162371
100 ul Each for $329.00
Add to cart
 
Description

Description

BTN2A1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

BTN2A1
Rabbit
Polyclonal
Unconjugated
11120
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to BTN2A1(butyrophilin, subfamily 2, member A1) The peptide sequence was selected from the N terminal of BTN2A1. Peptide sequence SVALVIHNITAQENGTYRCYFQEGRSYDEAILHLVVAGLGSKPLISMRGH.
Primary
IgG
BK14H9.1, BTF1BT2.1butyrophilin BTF1, butyrophilin subfamily 2 member A1, butyrophilin, subfamily 2, member A1, DJ3E1.1, FLJ36567
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit