Learn More
BTN3A3 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 158961AP
Catalog No. 50-173-0558
Add to cart
Description
Description
Plays a role in T-cell responses in the adaptive immune response.
Specifications
Specifications
BTN3A3
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00478
BTF3, BTN3A3
Rabbit
IgG
150 μL
-20°C
Primary
10384
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
BTN3A3
Liquid
BTN3A3
BTN3A3 Fusion Protein Ag8711
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human