Learn More
BTNL8 Rabbit anti-Human, Alexa Fluor 350, Clone: 2187B, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB9359U100UG
Catalog No. FAB9359U100
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
BTNL8
2187B
Flow Cytometry 0.25-1 ug/10^6 cells
Q6UX41-1
BTN9.2, butyrophilin-like 8, butyrophilin-like protein 8, UNQ702/PRO1347
Human embryonic kidney cell line HEK-293-derived Human BTNL8 Gln18-Lys238 Accession # Q6UX41-1
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human BTNL8 in direct ELISAs and Western blots.
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Supernatant
Rabbit
IgG
100 ug
Primary
79908
Human