Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTNL8 Rabbit anti-Human, Alexa Fluor 405, Clone: 2187B, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB9359V100UG
This item is not returnable.
View return policy
Description
BTNL8 Monoclonal specifically detects BTNL8 in Human samples. It is validated for Flow Cytometry.Specifications
BTNL8 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
BTN9.2, butyrophilin-like 8, butyrophilin-like protein 8, UNQ702/PRO1347 | |
Human embryonic kidney cell line HEK-293-derived Human BTNL8 Gln18-Lys238 Accession # Q6UX41-1 | |
100 μg | |
79908 | |
Human | |
Supernatant |
Flow Cytometry | |
2187B | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q6UX41-1 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human BTNL8 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title