Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTX™ Electroporation Accessories: Cables, Connectors, and Adapters

Connect external devices (Genetrodes, Genepaddles, Microslides, Meander Fusion Chamber) to BTX electroporation generators

$204.00

Specifications

Length (English) Cable 10 ft.
Length (Metric) Cable 3.1m
View More Specs

Products 2
Catalog Number Mfr. No. Connection Type For Use With (Application) For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Connection Type For Use With (Application) For Use With (Equipment) Price Quantity & Availability  
BTX450216
View Documents
BTX™
450216
Banana plug to micrograbber Connect the genetrode, genepaddle, microslide, meander fusion chamber to the electroporation generator Electroporation generator
Each for $298.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
BTX450217
View Documents
BTX™
450217
Banana plug to banana plug Required to connect the flat electrode to the electroporation generator ECM 830, ECM 2001 generator
Each for $204.00
Add to cart
 
Description

Description

Specifications

Specifications

10 ft.
3.1m
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit