Learn More
BTX 96 Well Plate Handler
Designed to work with 96 well plates. BTX Harvard Apparatus 96 Well Plate Handler helps to create a safe and reliable working area for processing electroporation plates.
Catalog Number | Mfr. No. | Description | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Includes | Price | Quantity & Availability | ||||
BTX450400
|
BTX
450400 |
Model: HT 100 Plate Handler | Manual track switching, Connectors for standard pulse generators |
Each for $4,074.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
BTX450401
|
BTX
450401 |
Model: HT 200 Plate Handler | Auto sense track switching, Connectors for standard pulse generator |
Each for $6,483.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
Description
- Quickly optimize for better gene delivery
- Designed to work with 96 well plates
- Pulse switching technology integrated into package and gold plated contacts to mate with electroporation plate
- With an adapter to accommodate 25 well plates as well with our ECM 630 generator
- Prosess up to 96 wells in seconds