Learn More
BTX™ HT 25-,96-Well ECM 630 Electroporation Systems
High-throughput systems increase cell yields and viability
Manufacturer:  BTX™ 450423
Catalog No. BTX450423
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Includes: HT-100 Plate Handler with connectors for standard pulse generators, ECM generator as specified in ordering table
Description
Description
Quickly and efficiently optimize the electrical and biological parameters associated with electroporation. Fast, precise pulsing for efficient molecular delivery using BTX patented HT 96-well electroporation plates.
  • Perform multiple experiments in either 5 x 5, 25-well plates or 8 x 12, 96-well plates
  • Research 96 experimental parameters in minutes
  • Exponential decay wave, for molecular biology applications
  • Adapter enables use of 25-well plates in the HT-100 plate handler
Plate Handler
  • HT 96-well plate handlers (HT-100) accommodate both 25- and 96-well plates
  • Features integrated automatic pulse switching technology and gold-plated contacts to mate with electroporation plates
  • Designed for use in small spaces, such as laminar flow hoods
  • HT-100 handler operates with the ECM630 and uses manual track switching, ideal for laboratories doing bacterial and yeast transformations
Transfection of mammalian cells; transformation of bacteria and yeast; cDNA library screening; drug screening; siRNA
Order Info
HT-100 Plate Handle can also be purchased separately (Cat. No. BTXHT100), as well as additional electroporation plates (Cat. No. BTX45-0462).
Specifications
Specifications
HT-200 plate handler with connectors for standard pulse generators, ECM generator as specified in ordering table
Transfection of mammalian cell, transformation of bacteria and yeast, siRNA and cDNA library screening