Learn More
BTX HT96 Enhancer 3000 Electroporation/Monitoring Systems
Versatile systems combine rapid multi-well electroporation with customizable data monitoring and analysis for fast, accurate results
Specifications
For Use With (Application) | Transformation of bacteria and yeast, transfection of mammalian cells |
---|---|
Includes | ECM generator, HT-200 plate handler, two 96-well HT plates, enhancer 3000 monitoring system with communications module |
Includes:
- ECM Generator
- HT-200 Plate Handler
- Two 96-well HT plates
- Enhancer 3000 Monitoring System with communications module
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
BTX450472
|
BTX
450472 |
ECM 830 generator |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
BTX450471
|
BTX
450471 |
ECM 630 generator |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
Description
The HT-96 Multi-Well System uses 96-well plates for faster turnaround than traditional cuvets. Run multiple experiments simultaneously, or load each plate with the same sample for high throughput processing. The Enhancer 3000 System enables the user to observe waveform, confirm data, track results, and document key parameters for protocol optimization, troubleshooting, and routine quality control.
- Processes multiple samples in seconds for optimization of electroporation variables
- Complete user control over electroporation settings allows for a wide range of optimization flexibility
- Results are confirmed automatically
- Monitoring and documentation of waveforms and electroporation parameters lets you quickly determine potential problems
- Communications module offers standard printer and computer interface
Choice of Generators
- ECM 630, an exponential decay generator recommended for high throughput electroporation of bacterial and yeast samples
- ECM 830, a square wave generator, for biomolecular screening, transfection, and gene silencing with no cytotoxicity
Specifications
Transformation of bacteria and yeast, transfection of mammalian cells | |
ECM generator, HT-200 plate handler, two 96-well HT plates, enhancer 3000 monitoring system with communications module |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.