Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BU 224 hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

205437-64-5
C12H12ClN3
233.699
DDFHQXAQWZWRSQ-UHFFFAOYSA-N
2-(4, 5-Dihydroimidazol-2-yl)quinoline hydrochloride
11957470

Potent, selective I2 ligand. Putative antagonist

High affinity ligand for the imidazoline I2 binding site (Ki = 2.1 nM). Putative I2 antagonist; antagonizes the effects of imidazoline ligands on morphine antinociception. Produces ipsiversive rotational behavior in rats with a full 6-OHDA lesion of the nigrostriatal tract.
Quantity 10mg
SMILES C1CN=C(N1)C2=NC3=CC=CC=C3C=C2.Cl
IUPAC Name 2-(4,5-dihydro-1H-imidazol-2-yl)quinoline;hydrochloride
Formula Weight 233.7
Chemical Name or Material BU 224 hydrochloride
Grade >98%

Recommended Storage : Desiccate at Room Temperature