BU 226 hydrochloride, Tocris

Potent, highly selective I2 ligand

Manufacturer: R&D Systems 1091/10

Quantity 10mg

 View more versions of this product

Catalog No. 10-911-0

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C12H11N3·HCl
    Formula Weight 233.7
    Synonym 2-(4, 5-Dihydroimidazol-2-yl)isoquinoline hydrochloride
    Chemical Name or Material BU 226 hydrochloride

    Specifications

    Formula Weight 233.7
    Quantity 10mg
    Assay Percent Range >98%
    Molecular Formula C12H11N3·HCl
    CAS 1186195-56-1
    Synonym 2-(4, 5-Dihydroimidazol-2-yl)isoquinoline hydrochloride
    Recommended Storage Store at Room Temperature
    Chemical Name or Material BU 226 hydrochloride

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit