Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BU 239 hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

1217041-98-9
C11H11ClN4
234.687
NQMMIEICYUXMKP-UHFFFAOYSA-N
2-(4, 5-Dihydroimidazol-2-yl)quinoxaline hydrochloride
56846046

Potent, highly selective I2 ligand

High affinity ligand for the imidazoline I2 binding site.
Quantity 10mg
SMILES C1CN=C(N1)C2=NC3=CC=CC=C3N=C2.Cl
IUPAC Name 2-(4,5-dihydro-1H-imidazol-2-yl)quinoxaline;hydrochloride
Formula Weight 234.69
Chemical Name or Material BU 239 hydrochloride
Grade >98%

Recommended Storage : Desiccate at Room Temperature