Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bub3 Mouse anti-Human, Alexa Fluor™ 350, Clone: 2H6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP242680AF350
Description
Bub3 Monoclonal specifically detects Bub3 in Human samples. It is validated for Western Blot, ELISA.Specifications
Bub3 | |
2H6 | |
Western Blot, ELISA | |
Purified, Conjugated | |
BUB3 | |
Recombinant human BUB3 (1-328aa) purified from E. coli | |
Protein A purified | |
Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
Primary | |
9184.0 |
Western Blot, ELISA | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
BUB3 (budding uninhibited by benzimidazoles 3, yeast) homolog, BUB3 budding uninhibited by benzimidazoles 3 homolog, BUB3L, budding uninhibited by benomyl, budding uninhibited by benzimidazoles 3 homolog (yeast), hBUB3, mitotic checkpoint component, mitotic checkpoint protein BUB3 | |
Mouse | |
IgG2b Kappa | |
100 μL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title