Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bub3 Mouse anti-Human, Alexa Fluor™ 750, Clone: 2H6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242680AF750

Catalog No. NP242680A70


Add to cart

Description

Description

Bub3 Monoclonal specifically detects Bub3 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Bub3
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
BUB3
Recombinant human BUB3 (1-328aa) purified from E. coli
100 μL
Primary
Human
IgG2b κ
Western Blot, ELISA
2H6
Western Blot, ELISA
BUB3 (budding uninhibited by benzimidazoles 3, yeast) homolog, BUB3 budding uninhibited by benzimidazoles 3 homolog, BUB3L, budding uninhibited by benomyl, budding uninhibited by benzimidazoles 3 homolog (yeast), hBUB3, mitotic checkpoint component, mitotic checkpoint protein BUB3
Mouse
Protein A purified
Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
9184.0
Store at 4°C in the dark.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit