Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BubR1 Mouse anti-Human, PE/Cy5.5, Clone: OTI6E5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270314PECY55

Catalog No. N270314PEC5


Add to cart

Description

Description

BubR1 Monoclonal antibody specifically detects BubR1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

BubR1
OTI6E5
Flow Cytometry
Affinity Purified, Conjugated
BUB1B
Full-length protein expressed in 293T cell transfected with human BubR1 expression vector.
Immunogen affinity purified
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers
Primary
701.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
Bub1A, BUBR1budding uninhibited by benzimidazoles 1 (yeast homolog), beta, budding uninhibited by benzimidazoles 1 homolog beta (yeast), EC 2.7.11.1, hBUBR1, MAD3/BUB1-related protein kinase, MAD3LBUB1beta, Mitotic checkpoint kinase MAD3L, Protein SSK1, SSK1mitotic checkpoint serine/threonine-protein kinase BUB1 beta
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit