Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BubR1 Mouse anti-Human, PerCP, Clone: OTI6E5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270314PCP
Description
BubR1 Monoclonal specifically detects BubR1 in Human samples. It is validated for Flow Cytometry.Specifications
BubR1 | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
BUB1B | |
Full-length protein expressed in 293T cell transfected with human BubR1 expression vector. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
OTI6E5 | |
Flow Cytometry | |
Bub1A, BUBR1budding uninhibited by benzimidazoles 1 (yeast homolog), beta, budding uninhibited by benzimidazoles 1 homolog beta (yeast), EC 2.7.11.1, hBUBR1, MAD3/BUB1-related protein kinase, MAD3LBUB1beta, Mitotic checkpoint kinase MAD3L, Protein SSK1, SSK1mitotic checkpoint serine/threonine-protein kinase BUB1 beta | |
Mouse | |
Affinity Purified | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
701.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title