Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI 1500 mL Beaker Flask
SDP

Two-piece flask offers easy access to product (glass flask only)

Supplier:  BUCHI 034140

Catalog No. 05-404-317


Add to Cart

Description

Description

  • Flat bottom stands firmly on lab bench
  • Can be used with bumping or foaming materials on a Rotavapor, or on a lyophilizer to dry sample completely without transfer
  • Capacity: 1500mL
  • Working Volume: 500mL
Specifications

Specifications

Beaker Flask
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit