Learn More
BUCHI™ Condenser Unit for Syncore™ Polyvap Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  BUCHI™ 037690
 View more versions of this product
Catalog No. 05-404-365
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
For condensing solvent vapor
Condenser unit S
Vertical condenser, vacuum adapter, support rod and clamps, PFA vacuum hose and fittings, 1000mL receiving flask