Learn More
BUCHI™ Drying Flasks for Rotary Evaporators Available on GSA/VA Contract for Federal Government customers only.
Special shaped flasks for powder drying
Manufacturer:  BUCHI™ 000454
 View more versions of this product
Catalog No. 05-402-670
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
29.2/32
CE
Rotary evaporator
Standard Glass
60°C
2000mL
2000mL
Powder Drying Flask
Plastic and Glass
29.2/32