BUCHI™ EPDM O-Ring for Multivapor™ Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: BUCHI™ 049676
 View more versions of this product
Catalog No. 05-402-892
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Used to seal the two units of the vacuum cover
    Size 193.7 x 7mm
    Product Type O-Ring
  • Description & Specifications
Related Products
Product Content Feedback