Learn More
BUCHI Evaporating Flask and Accessories Available on GSA/VA Contract for Federal Government customers only.
Flasks made of borosilicate 3.3 glass which is resistant to chemicals and temperature changes of 180°C
$70.60 - $736.50
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
05-403-203
View Documents
BUCHI
046596
Evaporating Flask Each for $736.50
Add to cart
05-403-206 Available on GSA/VA Contract for Federal Government customers only.
View Documents
BUCHI
048618
Flask Support Each for $70.60
Add to cart
Description
Description
  • Flask ring made of non-sliding silicone
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.