Learn More
BUCHI™ Evaporating Flasks for Rotary Evaporators
Use these evaporating flasks with any rotary evaporator. Buchi™ Evaporating Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for the distillation of solvents.
Manufacturer:  BUCHI™ 000431
 View more versions of this product
Catalog No. 05-402-658
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
29.2/32
50mL
Evaporating Flask
Uncoated
60°C
50mL
Rotary evaporator
Standard Glass
Pear
29.2/32