BUCHI™ Glass Column MPLC Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: BUCHI™ 019677
 View more versions of this product
Catalog No. 05-402-809
  Check Availability
Add to Cart
    Specifications
    Stationary Phase Sepacore™ C-690
    Type MPLC Glass column
    Column Type Sepacore™ C-690 MPLC Glass column
    Length (Metric) 230mm
    Diameter (Metric) Inner 80mm
    Material Borosilicate Glass
    Packing Material Sepacore™ C-690
  • Description & Specifications
Related Products
Product Content Feedback