Learn More
BUCHI™ Glass Columns C-690 Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  BUCHI™ 044053
 View more versions of this product
Catalog No. 05-402-634
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Preparative separations together with the C-600 Sepacore™ line of chromatography separators
Borosilicate Glass
460mm
145psi
Glass column
100mm