BUCHI™ Glass Plunger Column Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: BUCHI™ 044063
 View more versions of this product
Catalog No. 05-402-640
  Check Availability
Add to Cart
    Specifications
    Type Glass plunger column
    Column Type HPLC
    Length (Metric) 460mm
    Diameter (Metric) Inner 36mm
    For Use With Chemically resistant biocompatible plunger column with integrated cooling jacket
    Material Glass
  • Description & Specifications
Related Products
Product Content Feedback