Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI™ Glass Prep Column Available on GSA/VA Contract for Federal Government customers only.

Manufacturer:  BUCHI™ 044035

 View more versions of this product

Catalog No. 05-402-620


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Up to 53mL
580psi
Glass prep column
100mm
Used in combination with the injection unit for loading dry samples or barely soluble samples up to 53mL
Glass
HPLC
26mm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit