BUCHI™ Large Capacity Evaporating Flasks Available on GSA/VA Contract for Federal Government customers only.
Standard glass flasks for use with various Rotavapor™ Evaporator models
Manufacturer: BUCHI™ 002608
 View more versions of this product
Catalog No. 05-402-715
  Check Availability
Add to Cart
    Specifications
    Volume (Metric) 6L
    Capacity (Metric) 10000mL
    For Use With (Equipment) R-151 and R-152 rotary evaporator
    Product Type Evaporating Flask
    Material Standard Glass
  • Description & Specifications
Related Products
Product Content Feedback