BUCHI™ Multivapor™ P-6 Parallel Evaporation Systems with C Condenser for 100-120V Available on GSA/VA Contract for Federal Government customers only.

Evaporate up to 6 samples on one compact unit

Manufacturer: BUCHI™ MP22099C22

 View more versions of this product

Catalog No. 05-404-702

  Check Availability
Add to Cart

Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.


    Specifications

    Product Type Multivapor™ Parallel Evaporation System With C Condenser
    Certifications CE, CSA, ISO I4000
    Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
    Length (English) 5.74 in.
    Length (Metric) 40cm
    Voltage 100/120V
    Width (Metric) 27cm
    Width (English) 10.6 in.
    Model P-6
    Maximum Speed Range 0 to 485rpm
    Height (English) 5.74 in.
    For Use With (Equipment) 2L receiving flask
    Height (Metric) 40cm
    Hertz 50/60Hz
    Weight (English) 46.3 lb.
    Weight (Metric) 21kg
    Capacity (Metric) 10 to 160mL
    For Use With (Application) High throughput labs requiring multiple sample processing
    Height (English) Footprint 15.7 in.
    Includes V-700 pump, V-855 controller, woulff bottle, secondary condenser
    Height (Metric) Footprint 40cm
    Length (English) Footprint 10.2 in.
    Length (Metric) Footprint 26cm
    Temperature Range (English) 203°F
    Wattage 800w
    Volume (Metric) 2000mL
    Electrical Requirements 100-120V 50/60Hz
    Temperature Range (Metric) 95°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit