Learn More
BUCHI™ Multivapor™ P-6 Parallel Evaporators w/S Condenser, V-700/V-801, w/o Adapters, Safety Shield Available on GSA/VA Contract for Federal Government customers only.
Evaporate up to 6 samples on one compact unit
Manufacturer:  BUCHI™ MP22199S11
 View more versions of this product
Catalog No. 05-404-718
  Check Availability
Add to cart
Alert:
Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Specifications Description & Specifications
Specifications
CE, CSA, ISO I4000
5.74 in.
50/60Hz
40cm
46.3 lb.
27cm
P-6
10 to 160mL
15.7 in.
40cm
10.2 in.
203°F
34 oz.
5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Multivapor™ Parallel Evaporation System With S Condenser
40cm
5.74 in.
100/120V
21kg
10.6 in.
95°C
High throughput labs requiring multiple sample processing
Safety shield, V-700 pump, V-801 controller, woulff bottle, 1L receiving flask
0 to 485rpm
26cm
800w
1000mL
100-120V 50/60Hz