BUCHI™ Multivapor™ P-6 Parallel Evaporators w/S Condenser, V-700/V-801, Woulff bottle Available on GSA/VA Contract for Federal Government customers only.

Evaporates up to 6 samples on one compact unit

Manufacturer: BUCHI™ MP22099S21

 View more versions of this product

Catalog No. 05-404-694

  N/A
Add to Cart

Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Product Type Multivapor™ Parallel Evaporation System With S Condenser
    Certifications CE, CSA, ISO I4000
    Height (Metric) 40cm
    Weight (English) 46.3 lb.
    Width (Metric) 27cm
    Maximum Speed Range 0 to 485rpm
    Volume (English) 67.5 oz.
    Hertz 50/60Hz
    Length (English) 5.74 in.
    Length (Metric) 40cm
    Voltage 100/120V
    Weight (Metric) 21kg
    Width (English) 10.6 in.
    Model P-6 with 2L Receiving Flask
    Temperature Range (Metric) 95°C
    Capacity (Metric) 10 to 160mL
    For Use With (Application) High throughput labs requiring multiple sample processing
    Height (English) Footprint 15.7 in.
    Includes V-700 pump, V-801 controller, woulff bottle
    Height (Metric) Footprint 40cm
    Length (English) Footprint 10.2 in.
    Length (Metric) Footprint 26cm
    Temperature Range (English) 203°F
    Wattage 800w
    Volume (Metric) 2000mL
    Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
    Electrical Requirements 100-120V 50/60Hz
    Height (English) 5.74 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit