BUCHI™ Multivapor™ P-6 Parallel Evaporators w/S Condenser, w/o Adapter, Pump, Controller for 220-240V Available on GSA/VA Contract for Federal Government customers only.

Evaporates up to 6 samples on one compact unit

Manufacturer: BUCHI™ MP21199S20

 View more versions of this product

Catalog No. 05-404-772

  Check Availability
Add to Cart

Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.


    Specifications

    Product Type Multivapor™ Parallel Evaporation System With S Condenser
    Certifications CE, CSA, ISO I4000
    Height (Metric) 40cm
    Length (Metric) 40cm
    Voltage 220/240V
    Weight (English) 46.3 lb.
    Weight (Metric) 21kg
    Width (Metric) 27cm
    Width (English) 10.6 in.
    Model P-6
    Temperature Range (Metric) 95°C
    Capacity (Metric) 10 to 160mL
    For Use With (Application) High throughput labs requiring multiple sample processing
    Height (English) Footprint 15.7 in.
    Includes Safety shield, 2L receiving flask
    Height (Metric) Footprint 40cm
    Maximum Speed Range 0 to 485rpm
    Length (English) Footprint 10.2 in.
    Length (Metric) Footprint 26cm
    Temperature Range (English) 203°F
    Wattage 800w
    Volume (English) 67.5 oz.
    Volume (Metric) 2000mL
    Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
    Electrical Requirements 220-240V 50/60Hz
    Height (English) 5.74 in.
    Hertz 50/60Hz
    Length (English) 5.74 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit